To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Brain natriuretic peptide
- Clonality:Polyclonal
- Gene ID:2304
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BNP
- Conjugation:Unconjugated
- ImmunoPrecipitation:Yes
- Size:100 μl
- Western blot:Yes
- Antigen synonyms:brain natriuretic peptide|BNP|NPPB|natriuretic peptide B
- Storage buffer:1XPBS, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.
- Storage temperature:Keep as concentrated solution. Aliquot and store at –20 °C or below. Avoid multiple freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human FOXE1. The exact sequence is proprietary.
- Purification:Purified by antigen-affinity chromatography.
- Cat. no.:89318-198
- Supplier No.:GTX100275
Spécifications
A propos de cet article
Rabbit Polyclonal antibody to FOXE1 (forkhead box E1 (thyroid transcription factor 2))
Suggested starting dilutions are as follows: WB: 1:500-1:3000, IP: 1:100-1:500. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.
Type: Primary
Antigen: BNP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human