Digital Gauss Meter
# de catalogue 470164-000
Fournisseur: OSAW INDUSTRIAL PRODUCTS SE
Spécifications
- Description:Digital Gauss Meter
- Cat. no.:470164-000
- Supplier No.:SEA20
Spécifications
A propos de cet article
A useful tool for measuring the magnetic field strength for experiments. The range of the probe is 200 and 2kG with a resolution of 1G at 0 - 200 G. The included probe is a hall probe, which is a flat style probe to fit into small gaps.
Customers Who Bought This Also Bought