Laboratory Cleaner
Supplier: Ward's Science
Certificates
About this item
This cleaner is specially formulated for use on laboratory surfaces.
- For Lab Surfaces
- Ideal for Stainless Steel, Fume Hood Sashes, and More
- Product Is Designed for Educational Settings
It's ideal for surfaces such as stainless steel, stone and composite bench tops, and fume hood sashes.
This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Caution: Caution:The cleaner is intended for use in the removal of stains and residues. It is not intended for use as a disinfectant.
Customers Who Bought This Also Bought