Hydrion® Single Roll Dispenser, pH 6.0 to 8.0, Micro Essential Laboratory®
60784-204
- Description:Short Range pH Paper
- Cat. no.:60784-204
- pH Range:6.0 - 8.0
- Supplier No.:345
This short-range Hydrion pH paper is excellent for measuring pH solutions around neutral pH 7.00.
- Provides clear bright single color matches at every .2 to .3 interval from pH 6.0-8.0
- The color chart has pH matches at [6.0][6.4][6.8][7.0][7.2][7.6][8.0]
: Each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart. Provides for approximately 1000 tests.