Dispensing Bottle, HDPE, Round, Menda
Fournisseur: Desco
Certificats
A propos de cet article
Dispensing bottle is made from high denisty polyethylene/amber glass.
- Durable
- Alcohol imprint for demarcation
- Lasting touch pump
Ideal for alcohol dispensing. Amber glass construction protects light and plastic sensitive fluids.
Customers Who Bought This Also Bought