Sharps Bank Waste Disposal Unit | Avantor
Sharps Bank Waste Disposal Unit
45.4 L (12 gal.)
Catalog # 10796-894
Supplier:   Heathrow Scientific
Fournisseur #:  SB1201
Sharps Bank Waste Disposal Unit
Catalog # 10796-894
Sharps Bank Waste Disposal Unit
Sharps Bank Waste Disposal Unit
45.4 L (12 gal.)
Price
320,46 $
Prix promotionnel pour les nouveaux clients
Log in pour la disponibilité
Case of 15 (1/Itm/Bx)
Product Details & Documents
These containers are designed for the disposal of contaminated items in the lab environment.
They feature 3 walled construction: Inner and outer carton provide puncture resistance; a 3 mil liner provides leak resistance. They have an extra wide opening for safe, convenient disposal and the two-flap lid configuration protects contents from spilling. The containers have a square bottom to reduce the chance of tipping and the two handle construction makes transporting these containers to central disposal convenient. Containers ship and store flat for space-saving storage.
Spécifications
  • Capacité:
    45.4 L (12 gal.)