Oil for Belt Driven Pumps, HyVac
Fournisseur: Avantor
Certificats
A propos de cet article
Ideal for belt driven vacuum pumps
- Low vapor pressure
- Maintains oil seal
- Lubricates pump thoroughly
- Heat transfer efficiently
They resists absorption of condensates and are also thermally stable.
Customers Who Bought This Also Bought