PTFE Standard Taper Inner Joint to Sanitary Adapter, Ace Glass Incorporated
Fournisseur: Ace Glass
Certificats
A propos de cet article
This transition adapter converts a sanitary flanged apparatus to a standard taper inner joint.
- [ST] inner joint
- Made of PTFE
Manufactured from virgin polytetrafluoroethylene.
Customers Who Bought This Also Bought