CleanMark® Cleanroom Labels, Sterilization Indicating, Lgi
Fournisseur: Lgi
Certificats
A propos de cet article
Chemical/abrasion cleanroom labels are recommended for use in critical environments. The labels withstand moisture, chemical exposure, and extreme temperatures without smearing, cracking, or fading while maintaining adhesion throughout the sterilization process.
- Withstand extreme hot or cold temperatures, harsh chemical solvents, and physical abrasion
- Special inks change color when exposed to specific sterilization procedures
- Sterile process indication with a thermal transfer printable label, eliminating the need for a separate label and indicating dots or tape
They can indicate sterilization processes such as autoclaving, may be pre-sterilized, can firmly adhere to or easily peel from challenging surfaces, and will not contaminate clean or sterile environments with particulates, outgassing, or microbes.
Ordering information: CleanMark® labels can be custom constructed to fit any size, and can be used with either direct thermal or thermal transfer printers.
Customers Who Bought This Also Bought