Spécifications
- Description:Chemical Reactions Curriculum Learning Module
- Cat. no.:470163-832
- Supplier No.:74-6924
Spécifications
A propos de cet article
A Great Classroom Solution
- Includes detailed, standards-based coverage of:
- Changes of Matter
- Chemical Reactions
- Formulas and Equations
- Balancing Equations
- Types of Chemical Reactions
- Energy in Chemical Reactions
- Exothermic and Endothermic Reactions
- Chemical Reaction Rates and Catalysts
NewPath’s Chemical Reactions Curriculum Learning Module provides a complete set of visual, digital and hands-on resources designed to support teacher instruction and provide students with the skills to master key concepts on Chemical Reactions.
Each module includes:
1 Flip Chart Set with 10 double-side, laminated charts including a graphic overview and “write-on/wipe-off” activities
1 Student Learning Guide featuring self-directed readings, illustrated explanations, assessment questions and a lab investigation
1 Vocabulary Review Card Set
1 Curriculum Mastery Game (4 players)
1 Interactive Multimedia Lesson containing animated tutorials, interactive activities, visual resources, illustrated narrated glossary, virtual labs, and assessments