ContactUsLinkComponent
VWR® Low Form Griffin Beakers
VWR® Low Form Griffin Beakers
# de catalogue 10754-950
Fournisseur:  VWR International
VWR® Low Form Griffin Beakers
VWR® Low Form Griffin Beakers
# de catalogue 10754-950
Fournisseur:  VWR International
Log in for availability
Select an Option
150 ml (5 oz.)
Select an Option
20 - 140 ml
Select an Option
57.5 mm
Select an Option
150 ml
Select an Option
81 mm (3.19")
Select an Option
10754-950
A propos de cet article
These low form Griffin beakers are made from high quality Type 1, Class A, 33 expansion borosilicate glass.
  • Double capacity scale
  • Uniform wall thickness
  • Very good chemical resistance
  • High temperature resistance
  • Meets ASTM E960, E960-93
They feature a thick, slightly flared, beaded top with a spout designed to have excellent pouring characteristics. They are printed with white graduated scale and a durable marking area.
Customers Who Bought This Also Bought