Finntip®, Special Pipette Tips, Thermo Scientific
# de catalogue 14233-396
Fournisseur: Thermo Fisher Scientific
Spécifications
- Volume:100 - 1000 µL
- Tip Color:Blue
- Packed:5 Racks, 96 each
- Sterile:+
- Supplier No.:9401423
Spécifications
A propos de cet article
Finntip® Extended are manufactured from high quality polypropylene.
- Autoclavable
These long tapered tip especially adapted for deep tubes and narrow-necked bottles.
Customers Who Bought This Also Bought