Acids and Bases Kit
# de catalogue 470163-078
Fournisseur: Ward's Science
Spécifications
- Description:Acids and Bases Kit
- No. of Lab Groups:15
- Cat. no.:470163-078
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:IS8006
Spécifications
A propos de cet article
Use Standardization Techniques to Determine Molarity
- Classic AP Chemistry Experiment
- Instructions Included
A solution of sodium hydroxide which is approximately 0.01M will be standardized with a solid acid, potassium hydrogen phthalate, to determine the exact Molarity of the base. The standardized base will then be titrated against a monoprotic unknown acid to determine the molarity of the acid. A pH indicator and/or a pH meter will be used to determine the equivalence point.
Ordering information: Includes Hydrochloric Acid, Sodium Hydroxide, Potassium Hydrogen Phthalate, and Phenolphthalein.
Customers Who Bought This Also Bought