Ward's® Half Life Prop Radioactive Element
# de catalogue 470023-826
Fournisseur: Avantor
Spécifications
- Description:Wards Half Life Prop Radioactive Element
- No. of Lab Groups:15
- Cat. no.:470023-826
- Supplier No.:WARD470023-826
Spécifications
A propos de cet article
This is a great experiment to study half life and radioacticve elements in the classroom!
- Includes enough materials for 15 setups, a teacher's guide, and student copymasters
Radioisotopes have been found to be useful in the fields of geology, medical diagnosis and treatment, metallurgy, agriculture, and scientific research. This lab allows students to see a demonstration of nuclear decay using a model of uranium-238. By graphing their data, they better understand the reason for radioactive decay and the concepts of subatomic particles, half life, and more.
Customers Who Bought This Also Bought