Electron Deflection Tube Demonstration Series
# de catalogue 470162-714
Fournisseur: American 3B Scientific
Spécifications
- Description:Electron Deflection Tube Demonstration Series
- Cat. no.:470162-714
- Supplier No.:U19155
Spécifications
A propos de cet article
Highly evacuated electron tube with focusing electron gun and fluorescent screen inclined relative to the beam axis, so that the path of the beam can be seen and the effects of electric and magnetic fields can be studied. The electron beam can be deflected electrically in the electric field of the built-in plate capacitor, and magnetically by using the Helmholtz coil pair (not included). By adjusting the electric field so that it cancels the magnetic deflection, it is possible to determine the specific charge (e/m) and the velocity of the electrons.
Customers Who Bought This Also Bought