Electron Tube Holder Demonstration Series
# de catalogue 470162-710
Fournisseur: American 3B Scientific
Spécifications
- Description:Electron Tube Holder Demonstration Series
- Cat. no.:470162-710
- Supplier No.:U191001
Spécifications
A propos de cet article
Tube holder is made of robust plastic for holding D-series electron tubes. Stand has a wishbone, 360° rotating clamp made of heat-resistant plastic and two base holes for affixing the Teltron Helmholtz Coils. On rubber feet to prevent slipping.
Specifications:
Dimensions: approx. 230x175x320 mm³
Weight: approx. 1.5 kg
Customers Who Bought This Also Bought