3B Scientific® Whiteboard Optics Set
# de catalogue 470162-702
Fournisseur: American 3B Scientific
Spécifications
- Description:Whiteboard Optics Kit
- Cat. no.:470162-702
- Supplier No.:U14600
Spécifications
A propos de cet article
Easily Display Optics Principles
- Kit contains a mixture of reflectors, transparent and shadow objects
- Use your own light source or the multibeam projector
Set of optical components for use in conjunction with a single-beam lamp or multiple-beam lamp on a white board.
All components are lined with magnetic foil or furnished with a magnet holder and can be easily mounted and aligned on the white board. This apparatus permits a wide range of experiments demonstrating ray optics without the need for a dark room; handwritten notes can be added to provide a clearer explanation.
Customers Who Bought This Also Bought