Acid-Base Indicators
# de catalogue 470163-118
Fournisseur: Ward's Science
Spécifications
- Description:Acid-Base Indicators
- No. of Lab Groups:15
- Cat. no.:470163-118
- Includes:Teacher's Guide and Student Copymaster
- Supplier No.:IS8027
Spécifications
A propos de cet article
How Do Indicators Work?
- Classic AP Chemistry Experiment
- Instructions Included
pH is the measure of the concentration of hydrogen ions in a solution. As this concentration can extend over several orders of magnitude, it is convenient to express it by means of logarithms of base ten. Certain pigments change color with a change in pH. In this experiment we will extract pigments from various sources and determine if they are sensitive to changes in pH.
Ordering information: Includes Hydrochloric Acid, Sodium Hydroxide, Isopropyl Alcohol, and Acetone.
Customers Who Bought This Also Bought