Bernoulli Bag
# de catalogue 470163-924
Fournisseur: American Scientific
Spécifications
- Description:Bernoulli Bag
- Cat. no.:470163-924
- Supplier No.:7040-00
- No. of Parts:4
Spécifications
A propos de cet article
Explore Bernoulli's principle of air pressure.
- Comes with a teacher's guide
By placing the bag firmly to your mouth (like a balloon), it will take many breaths to blow it up. But if you hold it out in front of your mouth and blow, air pressure in the stream you produce is reduced, entrapping surrounding air to join in filling up the bag. So you can blow it up with a single breath! This is especially effective after your students have counted many of their own breaths in attempting to fill up the bag!
Customers Who Bought This Also Bought