Aluminium Weighing Dishes, Dyn-A-Med
Fournisseur: Dyn-A-Med
Certificats
A propos de cet article
Disposable aluminum foil dishes used for weighing or moisture testing.
- Used in wastewater, chemical, pharmaceutical, educational and dairy applications
Available with crimped sides, flat smooth bottom and a finger tab handle on the rim or totally smooth with no finger tab.
Customers Who Bought This Also Bought