Acid Rain Science Fair Kit
# de catalogue 470163-314
Fournisseur: Ward's Science
Spécifications
- Description:Acid Rain Science Fair Kit
- Cat. no.:470163-314
- Supplier No.:IS5032
- No. of Uses:100
Spécifications
A propos de cet article
Determine if You have Acid Rain in Your Area.
- Great field study
- Ideal for younger students
- All required chemicals are included
The Acid rain test kit is great to use in the classroom or for science fair projects to determine if you have acid rain in your area. Collect water samples and use a low range pH indicator to determine the pH of your water sample with a colorimetric comparison chart.
Ordering information: Includes Low Range pH Indicator, Test Tubes, Color Chart, and Instructions.
Caution: Great field study
Ideal for younger students
All required chemicals are included
Customers Who Bought This Also Bought