VWR® Volumetric Flasks with Glass Stopper, Class A
Fournisseur: VWR International
Certificats
A propos de cet article
These volumetric flasks are available unserialized or batch certified, and are calibrated at 20° Celsius.
- Graduated ring for measurement
- Durable white color printing
- Glass stopper fabrication
Made from ASTM E438 Type 1 Class A Borosilicate 33 expansion glass as per ASTM specification E237 and E288.
Customers Who Bought This Also Bought