Properties Of Matter: What Is Oobleck?
# de catalogue 470163-310
Fournisseur: Ward's Science
Spécifications
- Description:Properties of Matter: What is Oobleck?
- No. of Lab Groups:10
- Cat. no.:470163-310
- Supplier No.:IS1003
Spécifications
A propos de cet article
Hands On Science For Elementary Students!
- Ideal for grades K-4
- Introductory hands on activity
- Non-hazardous chemicals included
This kit has been designed to introduce young students (K-4) to the concepts of matter in a fun, safe, hands-on way. Students are given samples of both a solid and a liquid so that they can perform some simple experiments that demonstrate the differences. The students are then given a sample of “Our Friend Oobleck” (a non-Newtonian fluid) to use in the same experiments and are asked to determine if Oobleck is a solid or a liquid.
Ordering information: Includes Oobleck Powder, Solid Samples, Holding Trays, Testing Spatulas, and Teacher's Guide and Student Copymaster.
Customers Who Bought This Also Bought