Innovating Science® Acids and Bases Lab Activity
Fournisseur: Ward's Science
Certificats
A propos de cet article
In this activity, students make hypotheses regarding household items and then test them.
- Perfect Introduction to Acids and Bases
- Students Test Household Items
- Kit Features Extra Litmus Paper for More Experiments
- All Required Chemicals are Included
- Acid and Base Chemistry in Microscale
Students learn about acids and bases through a class discussion and then test a dilute acid and dilute base with neutral litmus paper to learn how the paper indicates something is acidic or basic.
Once students master that skill, they can move on to test some common household items to determine if they are acids or bases. Extra litmus paper is included to test more items either from home or around the school building for a greater understanding of acids and bases.
Certifications: Caution:CHOKING HAZARD This item has small parts and is not for children under three years.
Ordering information: This kit includes hydrochloric acid solution, sodium hydroxide solution, distilled water, vinegar, dilute ammonia, liquid soap, lemon juice, neutral litmus paper, spot plates, a teacher's guide, and a student copy master.
Delivery: Instructor's manual and SDS included.
Caution: Caution:CHOKING HAZARD This item has small parts and is not for children under three years.
Customers Who Bought This Also Bought