Separation of a Mixture: Physical Properties of Matter
# de catalogue 470163-288
Fournisseur: Ward's Science
Spécifications
- Description:Separation of a Mixture: Physical Properties of Matter
- No. of Lab Groups:6
- Cat. no.:470163-288
- Supplier No.:IS1000
Spécifications
A propos de cet article
Perfect Introduction to Physical Properties.
- Ideal for beginner scientists
- Separate a mixture based on only physical characteristics
- All chemicals required are included
After a class discussion of matter and the fact that matter may differ from other matter in a variety of ways, students will examine three different solids individually and observe some differences in physical properties. Sand, iron filings and salt will be observed and tested using water and magnets. Based on what they find individually they will come up with a method (as a class or as groups) to separate a mixture of all three.
Ordering information: Includes Sand, Salt, Iron Filings, 3 Part Mixture, Magnets, Plastic Cups, Magnifiers, and Teacher's Guide and Student Copymaster.
Customers Who Bought This Also Bought