Hunter Fire Assay Cupels, Bone Ash, Anachemia
Fournisseur: Avantor
Certificats
A propos de cet article
Hunter cupels have been a consistent mainstay in the assay industry for over 30 years
- Consistent industry mainstay
- For use in precious metal refinement
- Variety of sizes available
Used by precious metals refining and assay establishments worldwide, Hunter cupels are manufactured to exact and consistent standards using high purity bone ash. They are available in a variety of different sizes.
Customers Who Bought This Also Bought