Balance Pan Liners, Aluminum, Dyn-a-Med
Fournisseur: Avantor
Certificats
A propos de cet article
Compatible generic aluminum moisture pans for most moisture balances.
- Strong and lightweight
Multiple uses with liquid and solid samples including weighing, drying and moisture determination. The inexpensive disposable aluminum foil dishes with crimped sides and a flat smooth bottom are great for moisture testing, wastewater and many other applications.
Customers Who Bought This Also Bought