Stoichiometry In The Synthesis Of An Ionic Compound
# de catalogue 470163-088
Fournisseur: Ward's Science
Spécifications
- Description:Stoichiometry In The Synthesis of An Ionic Compound
- No. of Lab Groups:15
- Cat. no.:470163-088
- Supplier No.:IS8011
Spécifications
A propos de cet article
Create Potassium Alum Using Chemical Reactions
- Classic AP Chemistry Experiment
- Instructions Included
Many combinations of mono and trivalent cations yield crystals of the same stoichiometry and structure. The crystals are normally in the form of an octahedron. Sodium, potassium and ammonium ions are often the monovalent species whereas aluminum and chromium are examples of the trivalent ions. Potassium alum, KAl(SO4)2•12H2O, is the most common and is used in water purification, paper manufacturing and as a mordant in dyeing. In this experiment we will perform a series of chemical reactions which lead to the synthesis of the ionic compound potassium alum.
Includes:
Aluminum Metal Powder
Potassium Hydroxide Solution
Sulfuric Acid Solution
Ethanol/Water 50:50 V/V Solution
Caution: Classic AP Chemistry Experiment
Instructions Included
Customers Who Bought This Also Bought