Dissociation Constants of Weak Acids Kit
# de catalogue 470163-082
Fournisseur: Ward's Science
Spécifications
- Description:Dissociation Constants of Weak Acids Kit
- No. of Lab Groups:15
- Cat. no.:470163-082
- Supplier No.:IS8008
Spécifications
A propos de cet article
Its All About Equilibrium!
- Classic AP Chemistry Experiment
- Instructions Included
Students determine the equilibrium constant of three unknown acids and compare the data to known values to identify them.
Ordering information: Includes Benzoic Acid Crystals, Potassium Hydrogen Phthalate, Sodium Bisulfate, Sodium Hydroxide, and Phenolphthalein Solution.
Caution: Classic AP Chemistry Experiment
Instructions Included
Customers Who Bought This Also Bought