Quantofix Peroxide Test Strips, CTL Scientific Supply
Fournisseur: Macherey-Nagel
Certificats
A propos de cet article
Quantofix Peroxide for the semi-quantitative determination of hydrogen peroxide (H2O2) and peroxides in solutions.
Color changes with H2O2 from white to blue.
Ordering information: Packaged as a box of 100.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.