Spécifications
- Description:Formaldehyde Test Kit
- Storage:Room Temperature
- Cat. no.:10217-516
- Range:0-10-20-40-60-100-200 mg/L
- Supplier No.:91328
Spécifications
A propos de cet article
Quantofix Formaldehyde test strips and reagents for the semi-quantitative determination of formaldehyde in solutions
Quantofix Formaldehyde test kit has a gradation of scale: 0-10-20-40-60-100-200 mg/L HCHO beige to blue-violet.
Other aldehydes such as e.g. acetaldehyde or glutardialdehyde only react at higher concentrations, resulting in a discoloration which deviates from the color scale. Acetone does not react. Strong oxidizing and reducing agents cause low results.