ContactUsLinkComponent
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
# de catalogue 10217-516
Fournisseur:  Macherey-Nagel
Numéro CAS:  
Quantofix Formaldehyde Test Kit, CTL Scientific Supply
# de catalogue 10217-516
Fournisseur:  Macherey-Nagel
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Formaldehyde Test Kit
  • Storage:
    Room Temperature
  • Cat. no.:
    10217-516
  • Range:
    0-10-20-40-60-100-200 mg/L
  • Supplier No.:
    91328
Spécifications
A propos de cet article
Quantofix Formaldehyde test strips and reagents for the semi-quantitative determination of formaldehyde in solutions
Quantofix Formaldehyde test kit has a gradation of scale: 0-10-20-40-60-100-200 mg/L HCHO beige to blue-violet.
Other aldehydes such as e.g. acetaldehyde or glutardialdehyde only react at higher concentrations, resulting in a discoloration which deviates from the color scale. Acetone does not react. Strong oxidizing and reducing agents cause low results.
Ordering information: Packaged as a box of 100, strips and reagent.
Customers Who Bought This Also Bought