QUS Series Ultra Stack and Hang Bins, Quantum Storage Systems
Fournisseur: QUANTUM STORAGE SYSTEMS
Certificats
A propos de cet article
The QUS200 Series Ultra stack and hang bin is made from strong and durable injection molded plastic.
- Available in blue, yellow, red
- Autoclavable up to 250°F
- Resistant to extreme cold
- Large molded front label slot for ID label
Built-in rear hanger allows bins to hang from louvered panels of rails. Dividers maximize flexiblity and keep contents organized.
Customers Who Bought This Also Bought