ContactUsLinkComponent
Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Catalog # 10217-496
Supplier:  Macherey-Nagel
CAS Number:  
Quantofix Carbonate Hardness Test Strips, CTL Scientific Supply
Catalog # 10217-496
Supplier:  Macherey-Nagel
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Carbonate Hardness Test Strips
  • Storage:
    Room Temperature
  • Cat. no.:
    10217-496
  • Range:
    0-3-6-10-15-20 mg/L
  • Supplier No.:
    91323
Specifications
About this item
Quantofix Carbonate Hardness Test strips for the semi-quantitative determination of the carbonate hardness
Quantofix Carbonate Hardness Test strips have a gradation of scale: 0-3-6-10-15-20 mg/L light green to blue. Strong bases or other acid-consuming substances cause false results.
Carbonate hardness is that part of the total hardness resulting from the content of alkaline earth ions, which are equivalent to the carbonate and hydrogen carbonate ions present in the water. Thus the carbonate hardness is the portion of the total hardness which contributes to the formation of alkaline earth carbonates (boiler scale).
Ordering information: Packaged as a box of 100.
Customers Who Bought This Also Bought