Spécifications
- Description:Cryostat Lubrication Kit
- Cat. no.:10764-044
- Packaging:Metal Box
- Supplier No.:SPCLK04
Spécifications
A propos de cet article
This product is a quality alternative to Leica and is made from the highest quality, low-temperature synthetic lubricants.
- Special low temperature food grade synthetic lubricants
- Tested to temperatures down to –70 °C
- Lubricants are safe and odorless
- Light oil perfect for for cross bearing assemblies and any shaft roller bearings
Low temperature synthetic grease is perfect for mating surfaces of the cutting block, cutting head moving parts or the internal sliding mechanisms within the microtome.