Hareta Non-Stick Auto-Glass Coverslips, Springside Scientific
Fournisseur: SPRINGSIDE SCIENTIFIC, LLC.
Certificats
A propos de cet article
This specially treated coverglass is designed not to stick together resulting in fewer equipment jams and glass breakage.
- Packed in each small box to facilitate less loading events
- Specially coated and specifically designed for automatic coverslippers
Packed in an easy open vacuum-sealed case to eliminate moisture contamination.
Ordering information: Each case contains 10 small boxes and is foil vacuum-sealed. Each case contains 20 total ounces.
Customers Who Bought This Also Bought