Anti-PRKD3 Rabbit Polyclonal Antibody
# de catalogue 80083-818
Fournisseur: Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Protein Kinase D3
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human
- Antigen symbol:PRKD3
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- Size:400 µL
- Western blot:Yes
- Epitope:C-Terminal (875–890 aa)
- Cross adsorption:No
- Antigen synonyms:nu type|nPKC-NU|PKC-NU|PRKCN|nPKC-nu|Protein kinase C|PKD3|EPK2|Protein kinase EPK2
- Storage temperature:Cool
- Shipping temperature:Ice
- Cat. no.:80083-818
- Supplier No.:AP7025B
Spécifications
A propos de cet article
Type: Primary
Antigen: PRKD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal (875–890 aa)
Host: Rabbit
Isotype:
Reactivity: Human
Customers Who Bought This Also Bought