ContactUsLinkComponent
GxP Compliant Thermal Mapping Kit, ELPRO
GxP Compliant Thermal Mapping Kit, ELPRO
# de catalogue 10796-878
Fournisseur:  ELPRO SERVICES, INC.
MAPPING WAREHOUSE GMP
GxP Compliant Thermal Mapping Kit, ELPRO
# de catalogue 10796-878
Fournisseur:  ELPRO SERVICES, INC.
Spécifications
  • Description:
    GxP Temperature Mapping Kit
  • Cat. no.:
    10796-878
  • Supplier No.:
    810030
Spécifications
A propos de cet article
Save time and lower costs with ELPRO’s easy and efficient GMP-compliant mapping process.
  • No reports to compile
  • No special software to purchase
  • Requires minimal time and resources
  • Global service
  • No travel or on-site access required
As the global knowledge leader in environmental monitoring for over 30 years, ELPRO has helped countless customers improve their bottom line with our simple and efficient process for GMP-compliant mapping and reporting
Thermal Mapping in 4 Easy Steps: 1–Provide us a floor plan and specifications of your facility. 2–We provide data loggers with instructions for placement and activation. 3–You place the data loggers according to the mapping plan. 4–Download the PDF reports and email them to ELPRO for analysis.
ELPRO Facilities Mapping Kit covers up to a 10m(l) x 10m(w) x 5m(h) area and includes: Mapping Layout Plan based on the floor plan you provide; Includes project scope, acceptance criteria, test description, mapping layout, point placement rationale; Setup/Dismantle List and instructions for setting up the mapping loggers; Fifteen (15) temperature data loggers (Elpro LIBERO CB); Calibration Certificates; Detailed Analysis Mapping Report and one hour consultation with ELPRO Services validation engineer.
Applications include stability chambers, 2-8C cold room, pharmacy, truck/transport vehicle, cargo containers, small ambient rooms, clean rooms, laboratories.
Compare our turnkey remote mapping service with the competition. See why more and more companies are trusting ELPRO. GMP compliant thermal mapping reports for temperature controlled applications are simpler and easier with ELPRO.
Certifications: Global GMP compliance; EU PICS ANNEX 15 Compliance; IATA CEIV; and FDA Compliance.
Ordering information: To receive a formal quote for any project, contact your local Avantor representative today.
Customers Who Bought This Also Bought