To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:Streptavidin
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Antigen symbol:Streptavidin
- Conjugation:Unconjugated
- ELISA:Yes
- Size:10 mg
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS, pH 7.4, containing 0.02% sodium azide
- Storage temperature:The antibody is stable in lyophilized form if stored at -20 °C or below. The reconstituted antibody can be stored for 2-3 weeks at 2...8 °C. For long term storage, aliquot and store at -20 °C or below. Avoid repeated freezing and thawing cycles.
- Concentration:Reconstitute the lyophilized powder with deionized water (or equivalent) to an final concentration of 5 mg/ml
- Shipping temperature:Ice
- Immunogen:Purified recombinant streptavidin
- Tested applications:Dot blot
- Purification:Protein A/G affinity column
- Cat. no.:89494-988
- Packaging:Vial
- Supplier No.:A00621-10
Spécifications
A propos de cet article
GenScript Streptavidin Antibody is specific to streptavidin.
- High specificit
- High affinity
- High stability
GenScript offers highly specific monoclonal and polyclonal primary antibodies. These primary antibodies are validated for multiple applications, including Sandwich ELISA and western blot.
Type: Primary
Antigen: Streptavidin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: