ContactUsLinkComponent
VWR® Spouted Collection Containers
VWR® Spouted Collection Containers
# de catalogue 15704-047
Fournisseur:  VWR International
VWR* Sample Container
VWR® Spouted Collection Containers
# de catalogue 15704-047
Fournisseur:  VWR International
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Capacity:
    237 ml (8 oz.)
  • Cat. no.:
    15704-047
Spécifications
A propos de cet article
These 237 mL (8 oz.) containers are manufactured out of polystyrene with excellent clarity.
  • Graduated markings
  • Polystyrene construction
  • Convenient pour spout
  • Superior clarity
Convenient pour spout for simplified specimen retrieval. Each container is graduated in 10 mL (cc) and 0.5 oz. increments up to 6.5 oz.
Customers Who Bought This Also Bought