ContactUsLinkComponent
Hydrion® pH Strips, Micro Essential Laboratory
Hydrion® pH Strips, Micro Essential Laboratory
# de catalogue 62000-402
Fournisseur:  Micro Essential
Hydrion® pH Strips, Micro Essential Laboratory
# de catalogue 62000-402
Fournisseur:  Micro Essential
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Hydrion® pH Strips
  • Cat. no.:
    62000-402
  • pH Range:
    pH 1 - 12
  • Supplier No.:
    165-1-12
Spécifications
A propos de cet article
A direct reading pH paper with the widest range available in a single paper
A simple, single color match for all pH values. Distinct and easily matched color separations. Accurate, reproducible readings with an immediate response
Ordering information: Each carton contains 10 vials. Each vial contains 100 strips.
Customers Who Bought This Also Bought