WebSeal™ 384-Well Non-Coated Microtiter and Mid-well Plates
Supplier: Thermo Scientific
Certificates
About this item
WebSeal non-coated, plastic microplates have excellent solvent resistance and low background noise
Made of high-quality, GC-tested polypropylene. Ideal for pharmaceutical and industrial applications, as well as high-throughput screening, sample collection and storage, combinatorial chemistry and liquid phase assays.
Frequently Bought Together