Beaker-Style Collection Cup with Pour Spout, Therapak®
Catalogue # 10497-260
Fournisseur: Therapak, LLC
Spécifications
- Capacity:90 ml
- Description:Beaker Style Collection Cup
- Cat. no.:10497-260
- Supplier No.:74848
Spécifications
À propos de cet article
This miniature cup offers convenient collection and pour-off into urinalysis tubes or transport vials.
- Compact design
- Pour spout for easy transfer
- Available with temperature strip
The cup is graduated to 90 mL and can be stacked when being stored prior to collection.