Duramark® Thermoelectric Cooler/Warmers, Therapak®
Supplier: Therapak, LLC
Certificates
About this item
These products provide an ‘active’ solution for the transport of laboratory medical specimens.
Optional equipment includes a Battery Saver Switch which protects the car’s battery from premature draining as well as an AC Power Adapter. This adapter allows the unit to be used in any 110/120 volt AC wall outlet.
Using the 12-volt power plug in a vehicle, each unit will cool to approximately 40°F lower than ambient temperature or heat to around 135°F.
Ordering information: Custom temperature options are available. Contact your Avantor sales representative for more information.
Customers Who Bought This Also Bought