Large Screw Top Pathology Containers, Azer Scientific
Fournisseur: Azer Scientific Inc.
Certificats
A propos de cet article
Large screw top pathology containers facilitate secured specimen transport.
- High-Quality, High-Density Polyethylene
- Heavy-Duty Screw Cap Lid
- Moisture and Tamper-Resistant
- Histology-Reagent Resistant
Manufactured from high-quality, clarified polypropylene, these pathology containers are resistant to histology reagents.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.