Spécifications
- Description:Ts526 glass slide flap thermal transfer label kit (5 rolls, 1 ribbon, 1 cleaning wipe)
- Cat. no.:10769-994
- Packaging:2500 labels/kit
- Dimensions:0.875×0.875"
- Supplier No.:1020384
Spécifications
A propos de cet article
Glass Slide thermal transfer label kit is designed for use with glass slides exposed to chemical processing.
FLAP label technology (Flexible Lamination After Printing) protects images during staining and other harsh chemical processes.
They are capable of holding up to direct immersion in xylene, DMSO, methanol, isopropyl alcohol, and staining. Labels are kitted with matched ribbon and label stock to ensure a solution durable for virtually any process and environment.