Spécifications
- Description:Ts955 glass slide thermal transfer label kit
- Cat. no.:10769-992
- Packaging:28000 labels/kit
- Dimensions:22.2×22.4 mm (0.88×0.88")
- Supplier No.:1004781-1
Spécifications
A propos de cet article
Glass Slide thermal transfer label kit is designed for use with glass slides exposed to chemical processing.
They are capable of holding up to direct immersion in xylene, DMSO, methanol, isopropyl alcohol, and staining. Labels are kitted with matched ribbon and label stock to ensure a solution durable for virtually any process and environment.