To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:BMP/Retinoic Acid Inducible Neural Specific 1
- Clonality:Monoclonal
- Clone:2A10E3
- Host:Mouse
- Isotype:IgG2c
- Reactivity:Pig,Human,Mouse
- Antigen symbol:BRINP1
- Conjugation:Unconjugated
- ELISA:Yes
- Size:150 µL
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:DBCCR1|FAM5A|BC1
- Storage buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage temperature:–20 °C
- Immunogen:Recombinant Protein
- Purification:Protein A purification
- Cat. no.:10753-346
- Supplier No.:60336-1-IG
Spécifications
A propos de cet article
The DBC1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human DBC1. This antibody recognizes human, pig, mouse antigen. The DBC1 antibody has been validated for the following applications: ELISA, WB analysis.
Type: Primary
Antigen: DBC1
Clonality: Monoclonal
Clone: 2A10E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2c
Reactivity: Human, Pig, Mouse