To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Antibody type:Primary
- Antigen name:ButyrylCholinesterase
- Clonality:Polyclonal
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Antigen symbol:BCHE
- Conjugation:Unconjugated
- ELISA:Yes
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:150 µL
- Western blot:Yes
- Cross adsorption:No
- Antigen synonyms:Acylcholine acylhydrolase|CHE1|Cholinesterase|Choline esterase II|BCHE|Butyrylcholine esterase|Pseudocholinesterase
- Storage buffer:PBS with 0.02% Thimerosal, 50% Glycerol, pH 7.3.
- Storage temperature:–20 °C
- Immunogen:Fusion Protein
- Purification:Antigen Affinity purified
- Cat. no.:10273-138
- Supplier No.:23854-1-AP
Spécifications
A propos de cet article
The BCHE antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human BCHE. This antibody recognizes human antigen. The BCHE antibody has been validated for the following applications: ELISA, WB, IHC, IF analysis.
Type: Primary
Antigen: BCHE
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human