Transform, Nitrile Exam Gloves, Supermax Healthcare Canada
Fournisseur: SUPERMAX HEALTHCARE CANADA CA
Certificats
A propos de cet article
Optimal stretchability, just like latex.
- Beaded cuff snugs wrist, eliminates roll-up, minimizes entry of liquids
- Long-lasting comfort, durability and decreased hand
- Textured fingertips for precision gripping fatigue
- Hypoallergenic
- Protect hands from biohazards and a broad range of chemicals
Certifications: ISO 9001: 2000 certified by SGS (UK) and SIRIM (Malaysia)
ISO 13485: 2003 certified by SGS (UK)
CMDCAS ISO 13485 Certification
ISO 13485: 2003 certified by SGS (UK)
CMDCAS ISO 13485 Certification
Ordering information: Packaged 100 gloves per dispenser box.
Spécifications
- Catalog No:
- CA10500-564
- CA10500-566
- CA10500-568
- CA10500-560
- CA10500-562
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finger thickness:
- 0.10 mm
- 0.10 mm
- 0.10 mm
- 0.10 mm
- 0.10 mm
- Palm thickness:
- 0.07 mm
- 0.07 mm
- 0.07 mm
- 0.07 mm
- 0.07 mm
- ASTM No.:
- D6319:00a, EN 455 Part 1
- D6319:00a, EN 455 Part 1
- D6319:00a, EN 455 Part 1
- D6319:00a, EN 455 Part 1
- D6319:00a, EN 455 Part 1