Hydrocarbon Soluble Siliconizing Fluid
Fournisseur: Thermo Scientific
Certificats
A propos de cet article
Ideal for siliconizing glass surfaces.
- Short chain, clear polymeric silicone fluid consists primarily of dichlorooctamethyltetrasiloxane and is soluble in organic solvents
- Excellent for modifying metals, certain plastics, ceramics and fiber optics
When applied to glass, quartz or similar products, the unhydrolyzed chlorines present on the chain react with surface silanols to form a neutral, hydrophobic and tiightly bonded film over the entire surface.
Hydrocarbon soluble siliconizing fluid is acidic and care should be taken to avoid corrosion of metal that comes into contact with the liquid. The fluid is acidic only during application; after application, the surface is neutral.
Standard components: 1,7-Dichloro-1,1,3,3,5,5,7,7-Octamethyltetrasiloxan
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought